siRNA Details
Gene Symbol: Tomm34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044244.2 | NP_001037709.1 | 6, 7 | 837 | ||
XM_006235585.2 | XP_006235647.1 | 6, 7 | 1112 | |||
GenBank mRNA | BC105768.1 | - | 6, 7 | 838 | 7930355 |
