siRNA Details
Gene Symbol: Slc35c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107803.1 | NP_001101273.1 | 10 | 1365 | ||
XM_006235588.2 | XP_006235650.1 | 11 | 2239 | |||
XM_006235590.2 | XP_006235652.1 | 11 | 2114 | |||
XM_008762504.1 | XP_008760726.1 | 11 | 2124 | |||
XM_008762505.1 | XP_008760727.1 | 10 | 3043 | |||
GenBank mRNA | BC087085.1 | - | 10 | 2089 | ||
BC166837.1 | - | 11 | 1408 | 9027110 | ||
FQ233415.1 | - | 11 | 1466 |
