siRNA Details
Gene Symbol: Sulf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034927.1 | NP_001030099.1 | 18 | 3102 | ||
XM_006235595.2 | XP_006235657.1 | 18 | 3169 | |||
XM_006235596.2 | XP_006235658.1 | 18 | 2700 | |||
XM_008762506.1 | XP_008760728.1 | 18 | 2750 | |||
GenBank mRNA | AY742216.1 | - | 18 | 3102 | ||
BC091377.1 | - | 8 | 881 | |||
BC166410.1 | - | 18 | 2581 | 9026145 |
