siRNA Details
Gene Symbol: Fam217b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134551.1 | NP_001128023.1 | 3 | 954 | ||
XM_006235833.2 | XP_006235895.1 | 3 | 874 | |||
XM_006235834.2 | XP_006235896.1 | 4 | 962 | |||
XM_008762508.1 | XP_008760730.1 | 2 | 1210 |
