siRNA Details
Gene Symbol: Abhd16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014052.2 | NP_001014074.1 | 1 | 1578 | ||
GenBank mRNA | BC083664.1 | - | 1 | 1579 | 7135476 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Abhd16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014052.2 | NP_001014074.1 | 1 | 1578 | ||
GenBank mRNA | BC083664.1 | - | 1 | 1579 | 7135476 |