siRNA Details
Gene Symbol: Gpr155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107811.1 | NP_001101281.1 | 5 | 1374 | ||
XM_006234354.2 | XP_006234416.1 | 6 | 1520 | |||
XM_006234355.2 | XP_006234417.1 | 6 | 1516 | |||
XM_006234356.2 | XP_006234418.1 | 6 | 1520 | |||
XM_006234357.2 | XP_006234419.1 | 6 | 1520 |
