siRNA Details
Gene Symbol: Scrn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013162.1 | NP_001013180.1 | 4, 5 | 593 | ||
XM_006234358.2 | XP_006234420.1 | 4, 5 | 630 | |||
XM_006234359.2 | XP_006234421.1 | 4, 5 | 632 | |||
XM_006234360.2 | XP_006234422.1 | 4, 5 | 632 | |||
GenBank mRNA | BC088234.1 | - | 4, 5 | 593 | 7324882 |
