siRNA Details
Gene Symbol: Stkld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107822.1 | NP_001101292.1 | 14 | 1357 | ||
XM_006233785.2 | XP_006233847.1 | 14 | 1568 | |||
XM_008761643.1 | XP_008759865.1 | 14 | 1568 | |||
XM_008761644.1 | XP_008759866.1 | 14 | 1568 | |||
GenBank mRNA | BC161886.1 | - | 14 | 1340 | 7133966 |
