siRNA Details
Gene Symbol: Usp20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107827.3 | NP_001101297.1 | 15 | 1808 | ||
XM_006233916.2 | XP_006233978.1 | 15 | 2943 | |||
XM_008761666.1 | XP_008759888.1 | 13 | 1917 | |||
XM_008761667.1 | XP_008759889.1 | 13 | 1875 |
