siRNA Details
Gene Symbol: Fbxw2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107835.2 | NP_001101305.1 | 7 | 1476 | ||
XM_006233976.1 | XP_006234038.1 | 7 | 1610 | |||
XM_006233977.2 | XP_006234039.1 | 8 | 1526 | |||
XM_006233980.2 | XP_006234042.1 | 8 | 1434 | |||
XM_008761750.1 | XP_008759972.1 | 8 | 3085 | |||
XM_008761751.1 | XP_008759973.1 | 9 | 1691 | |||
XM_008761752.1 | XP_008759974.1 | 7 | 1412 | |||
GenBank mRNA | BC166990.1 | - | 7 | 1476 | 7110795 |
