siRNA Details
Gene Symbol: Rc3h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107839.1 | NP_001101309.1 | 13, 14 | 2476 | ||
XM_008761754.1 | XP_008759976.1 | 13, 14 | 2577 | |||
XM_008761755.1 | XP_008759977.1 | 12, 13 | 2271 |
