siRNA Details
Gene Symbol: Cadps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729291.4 | XP_002729337.1 | 1, 2 | 574 | ||
XM_003749705.3 | XP_003749753.1 | 1, 2 | 574 | |||
XM_006236147.2 | XP_006236209.1 | 1, 2 | 574 | |||
XM_006236152.2 | XP_006236214.1 | 1, 2 | 574 | |||
XM_006236153.2 | XP_006236215.1 | 1, 2 | 574 | |||
XM_006236154.2 | XP_006236216.1 | 1, 2 | 574 | |||
XM_231528.8 | XP_231528.4 | 1, 2 | 574 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001252107, NM_153163, NM_001252106, NM_001252105, NM_001252108, and others.
Gene Ontology Categories:
