siRNA Details
Gene Symbol: Klhdc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017456.1 | NP_001017456.1 | 8 | 1016 | ||
XM_006236201.2 | XP_006236263.1 | 9 | 1172 | |||
GenBank mRNA | BC085351.1 | - | 8 | 1016 | 7115168 |
