siRNA Details
Gene Symbol: Klhdc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017456.1 | NP_001017456.1 | 5, 6 | 842 | ||
XM_006236201.2 | XP_006236263.1 | 6, 7 | 998 | |||
GenBank mRNA | BC085351.1 | - | 5, 6 | 842 | 7115168 |
