siRNA Details
Gene Symbol: Cnot4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037782.1 | NP_001032871.1 | 6 | 917 | ||
XM_006236271.2 | XP_006236333.1 | 6 | 935 | |||
XM_006236272.2 | XP_006236334.1 | 6 | 935 | |||
XM_006236273.2 | XP_006236335.1 | 6 | 933 | |||
XM_006236274.2 | XP_006236336.1 | 6 | 933 | |||
XM_008762777.1 | XP_008760999.1 | 6 | 937 | |||
GenBank mRNA | BC100152.1 | - | 6 | 917 | 7456520 |
