siRNA Details
Gene Symbol: RGD1563986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044701.1 | NP_001038166.1 | 2 | 351 | ||
XM_006236361.2 | XP_006236423.1 | 2 | 1765 | |||
GenBank mRNA | DQ836251.1 | - | 2 | 351 |
