siRNA Details
Gene Symbol: Ppm1k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107863.1 | NP_001101333.1 | 5 | 1084 | ||
XM_006236571.2 | XP_006236633.1 | 6 | 1415 | |||
GenBank mRNA | FQ211521.1 | - | 6 | 1207 |
