siRNA Details
Gene Symbol: Rmnd5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_232051.8 | XP_232051.4 | 4 | 535 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rmnd5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_232051.8 | XP_232051.4 | 4 | 535 |