siRNA Details
Gene Symbol: Rmnd5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_232051.8 | XP_232051.4 | 9 | 1228 | ||
GenBank mRNA | FQ231069.1 | - | 4 | 363 |

More Information
Other Species Targeted


UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rmnd5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_232051.8 | XP_232051.4 | 9 | 1228 | ||
GenBank mRNA | FQ231069.1 | - | 4 | 363 |