siRNA Details
Gene Symbol: Gcfc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134554.1 | NP_001128026.1 | 5 | 791 | ||
XM_006236702.2 | XP_006236764.1 | 6 | 972 | |||
XM_006236703.2 | XP_006236765.1 | 6 | 972 |
