siRNA Details
Gene Symbol: Dysf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107869.1 | NP_001101339.1 | 10 | 944 | ||
XM_006236758.2 | XP_006236820.1 | 13 | 1900 | |||
XM_006236760.2 | XP_006236822.1 | 13 | 1899 | |||
XM_006236761.2 | XP_006236823.1 | 13 | 1587 | |||
XM_006236764.2 | XP_006236826.1 | 13 | 1899 | |||
XM_006236765.2 | XP_006236827.1 | 13 | 1587 | |||
XM_006236766.2 | XP_006236828.1 | 12 | 1806 | |||
XM_006236767.2 | XP_006236829.1 | 12 | 1494 | |||
XM_006236768.2 | XP_006236830.1 | 13 | 1899 | |||
XM_006236769.2 | XP_006236831.1 | 13 | 1587 | |||
XM_006236770.2 | XP_006236832.1 | 12 | 1806 | |||
XM_006236771.2 | XP_006236833.1 | 12 | 1494 | |||
XM_006236772.2 | XP_006236834.1 | 12 | 1806 | |||
XM_006236773.2 | XP_006236835.1 | 12 | 1494 | |||
XM_006236774.2 | XP_006236836.1 | 12 | 1806 | |||
XM_006236775.2 | XP_006236837.1 | 12 | 1494 | |||
XM_008763067.1 | XP_008761289.1 | 13 | 1587 | |||
XM_008763068.1 | XP_008761290.1 | 10 | 1010 | |||
XM_008763069.1 | XP_008761291.1 | 13 | 1902 |
