siRNA Details
Gene Symbol: Dysf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107869.1 | NP_001101339.1 | 1 | 71 | ||
XM_006236758.2 | XP_006236820.1 | 1 | 775 | |||
XM_006236760.2 | XP_006236822.1 | 1 | 774 | |||
XM_006236764.2 | XP_006236826.1 | 1 | 774 | |||
XM_006236766.2 | XP_006236828.1 | 1 | 774 | |||
XM_006236768.2 | XP_006236830.1 | 1 | 774 | |||
XM_006236770.2 | XP_006236832.1 | 1 | 774 | |||
XM_006236772.2 | XP_006236834.1 | 1 | 774 | |||
XM_006236774.2 | XP_006236836.1 | 1 | 774 | |||
XM_008763069.1 | XP_008761291.1 | 1 | 777 |
