siRNA Details
Gene Symbol: Rbsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107875.1 | NP_001101345.1 | 12 | 2032 | ||
XM_006236908.2 | XP_006236970.1 | 12 | 2047 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rbsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107875.1 | NP_001101345.1 | 12 | 2032 | ||
XM_006236908.2 | XP_006236970.1 | 12 | 2047 |