siRNA Details
Gene Symbol: Prickle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107876.1 | NP_001101346.1 | 7 | 1079 | ||
XM_006236912.2 | XP_006236974.1 | 7 | 1718 | |||
XM_006236913.2 | XP_006236975.1 | 7 | 6028 | |||
XM_008763124.1 | XP_008761346.1 | 7 | 1719 | |||
XM_008763125.1 | XP_008761347.1 | 7 | 1057 |
