siRNA Details
Gene Symbol: Adamts9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107877.1 | NP_001101347.1 | 16 | 2670 | ||
XM_006236915.2 | XP_006236977.1 | 17 | 2808 | |||
XM_006236916.2 | XP_006236978.1 | 17 | 2808 | |||
XM_008763126.1 | XP_008761348.1 | 17 | 2808 |
