siRNA Details
Gene Symbol: Adipor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037979.1 | NP_001033068.1 | 1 | 147 | ||
XM_006237183.1 | XP_006237245.1 | 2 | 453 | |||
XM_006237184.2 | XP_006237246.1 | 2 | 324 | |||
XM_006237185.2 | XP_006237247.1 | 2 | 339 | |||
XM_006237186.2 | XP_006237248.1 | 2 | 468 | |||
GenBank mRNA | AY724514.1 | - | 2 | 407 | ||
DQ148392.1 | - | 1 | 147 | |||
FQ232508.1 | - | 2 | 423 |
