siRNA Details
Gene Symbol: Ccdc77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191752.1 | NP_001178681.1 | 7, 8 | 818 | ||
XM_006237236.2 | XP_006237298.1 | 8, 9 | 1064 | |||
XM_006237237.2 | XP_006237299.1 | 8, 9 | 1052 | |||
XM_006237238.1 | XP_006237300.1 | 9, 10 | 1107 | |||
XM_006237239.1 | XP_006237301.1 | 9, 10 | 1089 | |||
XM_006237240.2 | XP_006237302.1 | 8, 9 | 1975 | |||
XM_006237241.1 | XP_006237303.1 | 8, 9 | 910 | |||
XM_006237242.2 | XP_006237304.1 | 7, 8 | 1995 | |||
XM_006237243.2 | XP_006237305.1 | 7, 8 | 1921 | |||
XM_006237244.2 | XP_006237306.1 | 7, 8 | 818 | |||
XM_008763244.1 | XP_008761466.1 | 8, 9 | 1064 | |||
XM_008763245.1 | XP_008761467.1 | 8, 9 | 2049 | |||
XM_008763246.1 | XP_008761468.1 | 8, 9 | 905 |
