siRNA Details
Gene Symbol: Eps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237583.2 | XP_006237645.1 | 6 | 1323 | ||
XM_008763452.1 | XP_008761674.1 | 6 | 607 | |||
XM_232499.8 | XP_232499.6 | 5 | 906 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Eps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237583.2 | XP_006237645.1 | 6 | 1323 | ||
XM_008763452.1 | XP_008761674.1 | 6 | 607 | |||
XM_232499.8 | XP_232499.6 | 5 | 906 |