siRNA Details
Gene Symbol: Eps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237583.2 | XP_006237645.1 | 10, 11 | 1812 | ||
XM_008763452.1 | XP_008761674.1 | 10, 11 | 1096 | |||
XM_232499.8 | XP_232499.6 | 9, 10 | 1395 |
