siRNA Details
Gene Symbol: Cmas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009419.1 | NP_001009419.1 | 4 | 625 | ||
GenBank mRNA | BC087599.1 | - | 4 | 625 | 7313414 | |
FQ230721.1 | - | 3 | 240 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cmas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009419.1 | NP_001009419.1 | 4 | 625 | ||
GenBank mRNA | BC087599.1 | - | 4 | 625 | 7313414 | |
FQ230721.1 | - | 3 | 240 |