siRNA Details
Gene Symbol: Cmas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009419.1 | NP_001009419.1 | 5, 6 | 776 | ||
GenBank mRNA | BC087599.1 | - | 5, 6 | 776 | 7313414 | |
FQ230721.1 | - | 4, 5 | 391 |

More Information
Other Species Targeted

Gene Ontology Categories:
