siRNA Details
Gene Symbol: Tram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007701.1 | NP_001007702.1 | 305 | |||
GenBank mRNA | AY387055.1 | - | 188 | |||
BC083851.1 | - | 305 | 7123274 | |||
FQ223598.1 | - | 299 |
