siRNA Details
Gene Symbol: Cpa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107900.1 | NP_001101370.1 | 7 | 684 | ||
XM_008763496.1 | XP_008761718.1 | 7 | 748 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cpa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107900.1 | NP_001101370.1 | 7 | 684 | ||
XM_008763496.1 | XP_008761718.1 | 7 | 748 |