siRNA Details
Gene Symbol: Ubxn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107905.1 | NP_001101375.1 | 3 | 329 | ||
XM_006237837.2 | XP_006237899.1 | 3 | 347 | |||
XM_008763540.1 | XP_008761762.1 | 2 | 222 | |||
XM_008763541.1 | XP_008761763.1 | 3 | 347 |
