siRNA Details
Gene Symbol: Ubxn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107905.1 | NP_001101375.1 | 7, 8 | 868 | ||
XM_006237837.2 | XP_006237899.1 | 7, 8 | 886 | |||
XM_008763540.1 | XP_008761762.1 | 6, 7 | 761 |

More Information
Other Species Targeted

Gene Ontology Categories:
