siRNA Details
Gene Symbol: Fbxl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107919.1 | NP_001101389.1 | 8 | 1687 | ||
XM_006237942.1 | XP_006238004.1 | 8 | 1694 | |||
XM_006237943.2 | XP_006238005.1 | 5 | 1377 | |||
GenBank mRNA | BC161859.1 | - | 8 | 1694 | 7096047 |
