siRNA Details
Gene Symbol: Mms22l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135780.1 | NP_001129252.1 | 11, 12 | 1685 | ||
XM_006237954.2 | XP_006238016.1 | 11, 12 | 1737 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mms22l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135780.1 | NP_001129252.1 | 11, 12 | 1685 | ||
XM_006237954.2 | XP_006238016.1 | 11, 12 | 1737 |