siRNA Details
Gene Symbol: Casp8ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107921.1 | NP_001101391.1 | 7, 8 | 2869 | ||
XM_006237973.2 | XP_006238035.1 | 8, 9 | 2769 | |||
XM_006237974.2 | XP_006238036.1 | 8, 9 | 2945 | |||
XM_006237975.2 | XP_006238037.1 | 7, 8 | 2699 | |||
XM_008763620.1 | XP_008761842.1 | 7, 8 | 3863 |
