siRNA Details
Gene Symbol: Orc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025282.1 | NP_001020453.1 | 8 | 724 | ||
XM_006237995.2 | XP_006238057.1 | 8 | 732 | |||
XM_008763621.1 | XP_008761843.1 | 8 | 729 | |||
XM_008763622.1 | XP_008761844.1 | 8 | 732 | |||
GenBank mRNA | AB219142.1 | - | 8 | 724 |
