siRNA Details
Gene Symbol: Nol6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238112.2 | XP_006238174.1 | 21 | 3913 | ||
XM_008763646.1 | XP_008761868.1 | 21 | 3913 | |||
GenBank mRNA | EF688596.1 | - | 9 | 1052 |
