siRNA Details
Gene Symbol: Nol6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238112.2 | XP_006238174.1 | 17, 18 | 3373 | ||
XM_008763646.1 | XP_008761868.1 | 17, 18 | 3373 | |||
GenBank mRNA | EF688596.1 | - | 5, 6 | 512 |
