siRNA Details
Gene Symbol: Ubap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107928.1 | NP_001101398.1 | 9 | 828 | ||
XM_006238051.2 | XP_006238113.1 | 9 | 760 | |||
XM_006238052.2 | XP_006238114.1 | 8 | 724 | |||
XM_006238053.2 | XP_006238115.1 | 9 | 760 | |||
XM_006238054.2 | XP_006238116.1 | 8 | 724 | |||
XM_008763625.1 | XP_008761847.1 | 8 | 1266 |
