siRNA Details
Gene Symbol: RGD1310951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238214.2 | XP_006238276.1 | 2 | 1542 | ||
XM_006238215.2 | XP_006238277.1 | 2 | 786 | |||
XM_008763745.1 | XP_008761967.1 | 2 | 786 |
