siRNA Details
Gene Symbol: Dcaf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107935.1 | NP_001101405.1 | 7 | 1416 | ||
XM_006238066.2 | XP_006238128.1 | 6 | 1254 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dcaf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107935.1 | NP_001101405.1 | 7 | 1416 | ||
XM_006238066.2 | XP_006238128.1 | 6 | 1254 |