siRNA Details
Gene Symbol: Dcaf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107935.1 | NP_001101405.1 | 3 | 775 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_153167, AK170534, BC027317, AK083428, BC085101, and others.
