siRNA Details
Gene Symbol: Kif12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012102.2 | NP_001012102.2 | 7 | 682 | ||
XM_006238253.2 | XP_006238315.1 | 7 | 700 | |||
XM_006238254.2 | XP_006238316.1 | 6 | 617 | |||
GenBank mRNA | BC085798.1 | - | 7 | 683 |
