siRNA Details
Gene Symbol: Kif12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012102.2 | NP_001012102.2 | 4, 5 | 405 | ||
XM_006238253.2 | XP_006238315.1 | 4, 5 | 423 | |||
XM_006238254.2 | XP_006238316.1 | 3, 4 | 344 | |||
GenBank mRNA | BC085798.1 | - | 4, 5 | 406 |
