siRNA Details
Gene Symbol: Kank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107947.1 | NP_001101417.1 | 3 | 1482 | ||
XM_008763868.1 | XP_008762090.1 | 2 | 1477 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107947.1 | NP_001101417.1 | 3 | 1482 | ||
XM_008763868.1 | XP_008762090.1 | 2 | 1477 |