siRNA Details
Gene Symbol: Tmem255a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182822.3 | NP_877974.2 | 1, 2 | 250 | ||
XM_006257479.2 | XP_006257541.1 | 1, 2 | 250 | |||
GenBank mRNA | AY327410.1 | - | 1, 2 | 115 | ||
CB580315.1 | - | 1, 2 | 178 | |||
CB706462.1 | - | 1, 2 | 250 | |||
CB745635.1 | - | 1, 2 | 256 |

More Information
Other Species Targeted

