siRNA Details
Gene Symbol: Orc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177931.2 | NP_808792.1 | 16 | 2464 | ||
XM_006238515.2 | XP_006238577.1 | 17 | 2666 | |||
XM_006238516.2 | XP_006238578.1 | 18 | 2779 | |||
GenBank mRNA | AB105378.1 | - | 16 | 2464 |
